Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD14 Alexa Fluor 405-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 1 publication
Supplier: R&D Systems FAB3832V025
This item is not returnable.
View return policy
Description
CD14 Monoclonal specifically detects CD14 in Human samples. It is validated for Flow Cytometry.Specifications
CD14 | |
Monoclonal | |
Alexa Fluor 405 | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
CD14, CD14 antigen, CD14 molecule, monocyte differentiation antigen CD14, Myeloid cell-specific leucine-rich glycoprotein | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
929 | |
Human | |
Supernatant |
Flow Cytometry | |
134620 | |
Flow Cytometry 5 uL/10^6 cells | |
P08571 | |
CD14 | |
Chinese hamster ovary cell line CHO-derived recombinant human CD14 Thr20-Cys352 Accession # P08571 | |
25 Tests | |
Primary | |
Detects human CD14 in direct ELISAs and Western blots. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction