Learn More
CD14 Mouse anti-Human, Clone: 50040, R&D Systems™
Mouse Monoclonal Antibody has been used in 2 publications
Manufacturer:  R&D Systems MAB3833100
 View more versions of this product
Catalog No. MAB3833100
Add to cart
This item is not returnable. View return policy
Description
Description
CD14 Monoclonal specifically detects CD14 in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).
Specifications
Specifications
CD14
50040
ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
P08571
CD14, CD14 antigen, CD14 molecule, monocyte differentiation antigen CD14, Myeloid cell-specific leucine-rich glycoprotein
Mouse
IgG1
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human CD14 in sandwich ELISAs.
Human
ELISA
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS and NaCl with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Supernatant
CD14
Chinese hamster ovary cell line CHO-derived recombinant human CD14 Thr20-Cys352 Accession # P08571
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
929
Reconstitute at 0.5 mg/mL in sterile PBS.