Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD14 Mouse anti-Human, Clone: 50040, R&D Systems™

Mouse Monoclonal Antibody

$119.60 - $839.00

Specifications

Antigen CD14
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human CD14 Thr20-Cys352 Accession # P08571
Clone 50040
Applications ELISA
Conjugate Unconjugated
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB3833SP
View Documents
R&D Systems
MAB3833SP
25 ug
Each for $119.60
Add to cart
 
MAB3833100
View Documents
R&D Systems
MAB3833100
100 ug
Each for $377.00
Add to cart
 
MAB3833500
View Documents
R&D Systems
MAB3833500
500 ug
Each for $839.00
Add to cart
 
Description

Description

CD14 Monoclonal antibody MAB3833 Specifically detects CD14 in Human samples. It is validated for ELISA Capture
Specifications

Specifications

CD14
50040
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS and NaCl with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Mouse
IgG1
929
RUO
Chinese hamster ovary cell line CHO-derived recombinant human CD14 Thr20-Cys352 Accession # P08571
ELISA
Purified
P08571
Monoclonal
Primary
Detects human CD14 in sandwich ELISAs.
Protein A or G purified from hybridoma culture supernatant
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit