Learn More
anti-CD14, PerCP, Clone: 134620, R&D Systems™
Manufacturer:  R&D Systems FAB3832C100
 View more versions of this product
Catalog No. FAB3832C100
Add to cart
Specifications
Specifications
CD14
134620
Flow Cytometry 10 uL/10^6 cells
P08571
CD14, CD14 antigen, CD14 molecule, monocyte differentiation antigen CD14, Myeloid cell-specific leucine-rich glycoprotein
Mouse
IgG1
100 Tests
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Monoclonal
Detects human CD14 in direct ELISAs and Western blots.
Flow Cytometry
PerCP
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Purified
CD14
Chinese hamster ovary cell line CHO-derived recombinant human CD14 Thr20-Cys352 Accession # P08571
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
929
Human