Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD14 Chicken anti-Human, Polyclonal, Invitrogen™

Chicken Polyclonal Antibody

Manufacturer:  Invitrogen PA120845

Catalog No. PIPA120845


Add to cart

Description

Description

PA1-20845 detects CD14 in Human samples. PA1-20845 has been successfully used in Western Blot procedures. PA1-20845 immunogen corresponds to Amino acids 40-230 of Human CD14..

CD14 is a 55 kDa GPI-anchored glycoprotein that is constitutively expressed on the surface of mature monocytes, macrophages, and neutrophils. CD14 also serves as a multifunctional lipopolysaccharide receptor, and is released to the serum both as a secreted and enzymatically cleaved GPI-anchored form. CD14 binds lipopolysaccharide molecule in a reaction catalyzed by lipopolysaccharide-binding protein (LBP), an acute phase serum protein. The soluble sCD14 can discriminate slight structural differences between lipopolysaccharides and is important for neutralization of serum allochthonous lipopolysaccharides by reconstituted lipoprotein particles. Further, CD14 has been shown to bind apoptotic cells, and can affect allergic, inflammatory and infectious processes. Alternative splicing results in multiple transcript variants encoding the same CD14 isoform. Diseases associated with CD14 dysfunction include mycobacterium chelonae infection and Croup.
Specifications

Specifications

CD14
Polyclonal
Unconjugated
CD14
Liquid
CD14
Amino acids 40-230 of Human CD14.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
P08571
monocyte differentiation antigen CD14, myeloid cell-specific leucine-rich glycoprotein
Chicken
IgY
50 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
929
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit