Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD140b Mouse anti-Human, Unlabeled, Clone: 28, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610114

 View more versions of this product

Catalog No. BDB610114

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Platelet-derived growth factor (PDGF) is a potent mitogen for cells of esenchymal origin and exerts its effects by binding to the PDGF receptor (PDGFR), a transmembrane protein tyrosine kinase. Both PDGF and PDGFR consist of subunits that form homo- or heterodimers with varying specificities: PDGF-AA binds only to αα PDGFR, PDGF-AB binds to both αα and αβ PDGFR, and PDGF-BB binds to all three PDGFRs. Ligand binding induces phosphorylation and activation of the receptor and, in turn, initiation of an intracellular phosphorylation cascade. PDGFR localizes primarily to membrane invaginations termed caveolae, compartments that are enriched in several of its downstream effectors, including PI3 kinase, Shc, NCK, and PTP1D. Additionally, the β subunit of PDGFR has been implicated in cellular transformation. This effect is mediated by a C-terminal region of PDGFRβ and depends on pathways that involve either PLCβ or PI3 kinase.

Host Species: Mouse
Clone: 28
Isotype: IgG2b
Species Reactivity: Human
Immunogen: Human PDGF Receptor β aa. 1-187
Formula Weight [Chemical]: 180kDa

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

Specifications

Specifications

CD140b
28
Unconjugated
Murine
IgG2b
RUO
Primary
Human
Immunofluorescence, Immunohistochemistry, Western Blot
250μg/mL
Affinity Purified
Human PDGF Receptor β aa. 1-187
150μg
Immunology
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.