Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD142 Rat anti-Mouse, Clone: 355220, Invitrogen™

Rat Monoclonal Antibody

Manufacturer:  Invitrogen MA524102

Catalog No. PIMA524102


Add to cart

Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS.

This gene encodes coagulation factor III which is a cell surface glycoprotein. This factor enables cells to initiate the blood coagulation cascades, and it functions as the high-affinity receptor for the coagulation factor VII. The resulting complex provides a catalytic event that is responsible for initiation of the coagulation protease cascades by specific limited proteolysis. Unlike the other cofactors of these protease cascades, which circulate as nonfunctional precursors, this factor is a potent initiator that is fully functional when expressed on cell surfaces. There are 3 distinct domains of this factor: extracellular, transmembrane, and cytoplasmic. This protein is the only one in the coagulation pathway for which a congenital deficiency has not been described.
Specifications

Specifications

CD142
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
P20352
F3, Coagulation factor 3, TF, TFA, Coagulation Factor III
Rat
IgG2C
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
14066
Western Blot
355220
Unconjugated
F3
Lyophilized
F3
Mouse myeloma cell line NS0-derived recombit mouse Coagulation Factor III Ala29-Glu251
Protein A/G
RUO
Antibody
Monoclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit