Learn More
CD147 Mouse anti-Mammalia, Clone: CB43, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA135092
Catalog No. 01-676-932
Add to cart
Description
Description
CD147 (basigin, neurothelin, OX-47, 5A11, CE9, M6, EMMPRIN, extracellular matrix metalloproteinase inducer, TCSF, tumour cell-derived collagenase-stimulatory factor) is a 50-60 kDa glycoprotein with multiple glycosylated forms. CD147 is a plasma membrane protein that is important in spermatogenesis, embryo implantation, neural network formation, and tumor progression. CD147 is also a member of the immunoglobulin superfamily. The highest level of CD147 expression is on metabolically active cells, such as lymphoblasts, inflammatory cells, brown adipocytes and maligt tumor cells. CD147 has multiple functions, including facilitating of cell surface expression of monocarboxylate transporter proteins and extracellular matrix metalloproteinases, regulation of integrin functions, it plays roles in cell development and activation, fetal development or retinal function.
Specifications
Specifications
CD147
Monoclonal
Unconjugated
BSG
Liquid
BSG
Native leucocytes.
Purified
RUO
Antibody
Monoclonal
Mammalia
Immunohistochemistry (Frozen)
CB43
Antibody with no preservative
Q865R3, P17790, P35613, P18572, Q28740, P26453
5F7, CD147, EMMPRIN, M6, OK, TCSF, CD147 antigen, OK blood group antigen, basigin, collagenase stimulatory factor, extracellular matrix metalloproteinase inducer, leukocyte activation antigen M6, tumor cell-derived collagenase stimulatory factor
Mouse
IgM
100 μg
4° C
Primary
100009229, 12215, 25246, 508716, 682, 770363