Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CTLA-4 Mouse anti-Human, Clone: AS32, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA112205

Catalog No. 01-670-915


Add to cart

Description

Description

CD152 is is expressed on most T lymphocytes. The level of expression is activation-dependent.

MA1-12205 has been shown to block the interaction of soluble hCD152 with hCD80
Specifications

Specifications

CTLA-4
Monoclonal
1 mg/mL
PBS with 0.08% sodium azide
P16410
CELIAC3, CTLA-4, GRD4, GSE, ICOS, IDDM12, CD152 isoform, celiac disease 3, cytotoxic T lymphocyte associated antigen 4 short spliced form, cytotoxic T-lymphocyte antigen 4
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
1493
Neutralization
AS32
Unconjugated
CTLA4
Liquid
CTLA4
Recombit extracellular domain of human CD152
Protein A/G
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit