Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-CD155/PVR, Alexa Fluor 488, Clone: 690912, R&D Systems™
Rat Monoclonal Antibody
Supplier: R&D Systems FAB6909G
This item is not returnable.
View return policy
Description
CD155/PVR Monoclonal specifically detects CD155/PVR in Mouse samples. It is validated for Flow Cytometry.Specifications
CD155/PVR | |
Monoclonal | |
Alexa Fluor 488 | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
CD155, CD155 antigen, HVED, Necl-5, NECL5, nectin-like 5, Nectin-like protein 5, poliovirus receptor, PVR, PVS, PVSFLJ25946, Tage4 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
5817 | |
Mouse | |
Purified |
Flow Cytometry | |
690912 | |
Flow Cytometry 5 uL/10^6 cells | |
NP_081790 | |
PVR | |
Mouse myeloma cell line NS0-derived recombinant mouse CD155/PVR Asp29-Leu348 Accession # NP_081790 | |
100 Tests | |
Primary | |
Detects mouse CD155/PVR in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2a |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title