Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD157 Mouse anti-Human, PE, Clone: SY/11B5, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 564214

Catalog No. BDB564214


Add to cart

Description

Description

The SY/11B5 monoclonal antibody specifically binds to CD157 which is also known as BST-1 (Bone marrow stromal antigen 1), BP-3/IF-7, Mo5, ADP-ribosyl cyclase 2, and cADPr hydrolase 2. CD157 is a 40-46 kDa glycophosphatidylinositol-linked cell membrane glycoprotein. It is an ectoenzyme that has both cyclic ADP-ribose hydrolase and ADP-ribosyl cyclase activities. CD157 is expressed as a homodimer by a variety of cell types including bone marrow stromal cells, granulocytes, monocytes, macrophages, dendritic cells, endothelial cells. and B and T cell progenitors. In addition to its ectoenzyme activities, CD157 reportedly functions as a receptor involved in neutrophil and monocyte adhesion, transendothelial migration and diapedesis and in tumor cell migration.

Specifications

Specifications

CD157
SY/11B5
BST1; Bone marrow stromal antigen 1; ADP-ribosyl cyclase 2; cADPr hydrolase
Affinity Purified
Human CD157 Transfected Cell Line
50 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
PE
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Murine
IgG1, κ
RUO
Primary
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit