Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-CD160, APC, Clone: 688327, R&D Systems™
Mouse Monoclonal Antibody
Supplier: R&D Systems FAB6700A
This item is not returnable.
View return policy
Description
CD160 Monoclonal specifically detects CD160 in Human samples. It is validated for Flow Cytometry.Specifications
CD160 | |
Monoclonal | |
APC | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
BY55, BY55FLJ46513, CD160, CD160 antigenimmunoglobulin superfamily member, CD160 molecule, Natural killer cell receptor BY55, NK1, NK28 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
11126 | |
Human | |
Purified |
Flow Cytometry | |
688327 | |
Flow Cytometry 10 uL/10^6 cells | |
O95971 | |
CD160 | |
Chinese hamster ovary cell line CHO-derived recombinant human CD160 Ile27-Ser159 Accession # O95971 | |
100 Tests | |
Primary | |
Detects human CD160 in direct ELISAs. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse CD160 is observed. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2b |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title