Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CD161, Alexa Fluor 488, Clone: PK136, R&D Systems™

Specifications

Clone PK136
Host Species Mouse
Isotype IgG2a
Immunogen Mouse NK-1+ splenocytes and bone marrow cells
Antigen CD161/NK1.1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
FAB8319G100
View Documents
R&D Systems
FAB8319G100
100 Tests This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
FAB8319G025
View Documents
R&D Systems
FAB8319G025
25 Tests This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Specifications

Specifications

PK136
IgG2a
CD161/NK1.1
Purified
RUO
Detects mouse CD161/NK1.1 by flow cytometry
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Mouse
Mouse NK-1+ splenocytes and bone marrow cells
Monoclonal
Primary
Flow Cytometry
17059
Protein A or G purified from hybridoma culture supernatant
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit