Learn More
anti-CD161, Alexa Fluor 700, Clone: PK136, R&D Systems™
Manufacturer:  R&D Systems FAB8319N100
 View more versions of this product
Catalog No. FAB8319N100
Add to cart
Specifications
Specifications
CD161/NK1.1
PK136
Flow Cytometry 5 uL/10^6 cells
Purified
KLRB1C
Mouse NK-1+ splenocytes and bone marrow cells
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
17059
Mouse
Flow Cytometry
Alexa Fluor 700
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
CD161, CD161 antigen, CD161c, CLEC5BC-type lectin domain family 5 member B, HNKR-P1a, killer cell lectin-like receptor subfamily B member 1, killer cell lectin-like receptor subfamily B, member 1, Klrb1c, Ly55c, Ly-59, Natural killer cell surface protein P1A, NKR-P1, NKR-P1AMGC138614, NKRP1ANKR
Mouse
IgG2a
100 Tests
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Monoclonal
Detects mouse CD161/NK1.1 by flow cytometry