Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD166 Mouse anti-Human, Alexa Fluor 647, Clone: 3A6, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 564938

Catalog No. BDB564938


Add to cart

Description

Description

The 3A6 monoclonal antibody specifically binds to the activated leukocyte cell adhesion molecule (ALCAM), a 100-105 kDa transmembrane glycoprotein, also known as the CD6 ligand, CD166. It is composed of an immunoglobulin-like extracellular domain, a transmembrane region and a short cytoplasmic tail. ALCAM belongs to the Ig superfamily of proteins and is expressed on neurons, activated T cells, activated monocytes, epithelial cells and fibroblasts. CD166 plays an important role in mediating adhesion interactions between thymic epithelial cells and CD6+ cells during intrathymic T-cell development.

Specifications

Specifications

CD166
3A6
ALCAM; Activated leucocyte cell adhesion molecule; MEMD
Affinity purified
IgG1, κ
RUO
Primary
Human
Flow Cytometry
Alexa Fluor 647
Aqueous buffered solution containing BSA and ≤0.09% sodium azide
Murine
50 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit