Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RHAMM/CD168 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP131797
Description
RHAMM/CD168 Polyclonal specifically detects RHAMM/CD168 in Human samples. It is validated for Western Blot.Specifications
RHAMM/CD168 | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
HMMR | |
Recombinant protein encompassing a sequence within the center region of human RHAMM. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
3161 | |
Human | |
IgG |
Western Blot | |
0.39mg/mL | |
Western Blot 1:500-1:3000 | |
CD168, CD168 antigen, hyaluronan mediated motility receptor, hyaluronan-mediated motility receptor (RHAMM), IHABP, Intracellular hyaluronic acid-binding protein, MGC119494, Receptor for hyaluronan-mediated motility, RHAMMMGC119495 | |
Rabbit | |
84 kDa | |
0.1 mL | |
Primary | |
Tested in human. Not yet tested in any other species. | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction