Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD180 Rat anti-Mouse, Biotin, Clone: RP/14, Invitrogen™

Rat Monoclonal Antibody

Manufacturer:  Invitrogen MA140035

Catalog No. 01-676-491


Add to cart

Description

Description

MA1-40035 detects CD180 from mouse samples. MA1-40035 has been successfully used in flow cytometry applications. The MA1-40035 immunogen is mouse RP105/CD180.

Radioprotective 105 (RP105, CD180) is a 105 kDa type 1 transmembrane protein consisting of extracellular leucine-rich repeats (LLRs) and a short cytoplasmic tail. RP105, assigned as CD180, is similar to Drosophila Toll in the extracellular LLRs. Radioprotective 105 (RP105, CD180) expression is largely restricted to immune cells including CD19+ B cells and macrophages. Histological studies showed that RP105 is expressed mainly on mature B cells in mantle zones. Antibody-mediated (RP/14) cross-linking of RP105 induces resistance against irradiation-induced apoptosis, B-cell proliferation, and up-regulation of a costimulatory molecule B7.2, revealing RP105 as a potent regulator of B-cell activation. RP105 (CD180) has an important role in B-cell activation by LPS. It is important to note that RP105 associates with MD1 and form the cell surface receptor complex, RP105/MD-1. It belongs to the family of pathogen receptors, Toll-like receptors (TLR). RP105/MD1, by working in concert with TLR4, controls B cell recognition and signaling of lipopolysaccharide (LPS), a membrane constituent of Gram-negative bacteria. MD-1 seems to be requisite for efficient expression of RP105.
Specifications

Specifications

CD180
Monoclonal
0.1 mg/mL
PBS with 0.1% BSA and 0.02% sodium azide
Q62192
LY64, Ly78, CD180 antigen, lymphocyte antigen 64, lymphocyte antigen-64 radioprotective, radioprotective 105 kDa protein
Rat
IgG2a
50 μg
4° C
Primary
17079
Flow Cytometry, Functional Assay
RP/14
Biotin
Cd180
Liquid
CD180
Murine RP105/CD180.
Protein G
RUO
Antibody
Monoclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit