Learn More
CD181 Mouse anti-Human, PE, Clone: 5A12, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 555940
Description
Reacts with Interleukin-8 receptor A (IL-8RA), a G protein-linked seven transmembrane domain protein, expressed on monocytes and granulocytes. It is also expressed on a subset of T cells. Both IL-8RA and IL-8RB are high affinity receptors for IL-8. IL-8 is an inflammatory cytokine which induces chemotaxis of neutrophils, basophils and T lymphocytes. IL-8RA and IL-8RB were first clustered as CD128a and CD128b, respectively in the VIIth HLDA workshop,and then were re-clustered as CD181 and CD182, respectively in the VIIIth HLDA workshop.
This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Specifications
CD181 | |
Monoclonal | |
0.2mg/mL | |
CXCR1, IL-8RA, CD128a | |
Mouse | |
0.2 mg | |
Immunology | |
Human | |
IgG2b κ |
Flow Cytometry | |
5A12 | |
PE | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.