Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD184 Rat anti-Mouse, Brilliant Violet 510, Clone: 2B11/CXCR4, BD

Rat Monoclonal Antibody

Manufacturer:  BD Biosciences 563468

Catalog No. BDB563468


Add to cart

Description

Description

The 2B11/CXCR4 monoclonal antibody specifically binds to mouse CD184 which is also known as the CXC chemokine receptor, CXCR4. CXCR4 (previously known as Fusin and LESTR), is a seven-transmembrane, G-protein-coupled receptor. It is the specific receptor for the CXC chemokine, SDF-1/CXCL12. Mouse CXCR4 shows 91% homology at the amino acid level with human CXCR4. CXCR4 is widely expressed by hematopoietic and non-hematopoietic cell types including neutrophils, monocytes, T cells, B cells, CD34-positive progenitor cells, endothelial cells, neurons and astrocytes. Human CXCR4 is used by T-tropic HIV-1 as a co-receptor for viral entry. The mouse Cxcr4 gene has been mapped to chromosome 1.

Specifications

Specifications

CD184
2B11/CXCR4
Brilliant Violet 510
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Rat
IgG2b, κ
RUO
Primary
Murine
Flow Cytometry
0.2mg/mL
CXCR4, C-X-C chemokine receptor type 4; Fusin; LESTR; PB-CKR; Sdf1r
Affinity Purified
GST-NCXCR4 fusion protein
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit