Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCR4 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA112542

Catalog No. 01-674-785


Add to cart

Description

Description

A suggested positive control for this product is HeLa cell lysate.

Human immunodeficiency virus (HIV) and related viruses require coreceptors, in addition to T-cell antigen T4 (CD4), to infect target cells. Some G protein-coupled receptors including chemokine (C-C) receptor 5 (CCR5), chemokine (C-X-C) receptor 4 (CXCR4), CCR3, CCR2b, and CCR8 in the chemokine receptor family, and four new human molecules G-protein coupled receptor 15 (GPR15), CXCR6, GPR1, and chemokine (C-X3-C) receptor 1 (CX3CR1) were recently identified as HIV coreceptors. Among them, CXCR4 (fusin, LESTR, or HUMSTR) is a principal coreceptor for T-cell tropic strains of HIV-1 fusion and entry of human white blood cells. CXCR4 is also required for the infection by dual-tropic strains of HIV-1 and mediates CD-4 independent infection by HIV-2. The alpha-chemokine stromal cell-derived factor-1 (SDF-1) is the ligand for CXCR4 and prevents infection by T-tropic HIV-1. CXCR4 associates with the surface CD4-gp120 complex before HIV enters target cells. CXCR4 messenger RNA levels correlate with HIV-1 permissiveness in diverse human cell types. Some antibodies to CXCR4 block HIV-1 and HIV-2 fusion and infection of human target cells. The amino-terminal domain and the second extracellular loop of CXCR4 serve as HIV binding sites.
Specifications

Specifications

CXCR4
Polyclonal
Unconjugated
CXCR4
Liquid
CXCR4
Synthetic peptide corresponding to residues 182-196 of CXCR4 receptor.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Western Blot
0.5 mg/mL
PBS with 0.02% sodium azide
P61073, P70658
D2S201E, FB22, HM89, HSY3RR, LAP3, LCR1, LESTR, NPY3R, NPYR, NPYRL, NPYY3R, WHIM, C-X-C chemokine receptor type 4
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
12767, 7852
Documents
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit