Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD19, APC, clone: 1G9, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA112258

Catalog No. 01-670-954

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

CD19 is a member of the immunoglobulin superfamily and has two Ig like domains. The CD19 molecule is expressed on 100% of the peripheral B cells as defined by expression of kappa or lamda light chains. It appears to be expressed on myeloid leukemia cells, particularly those of monocytic lineage. Leukemia phenotype studies have demonstrated that the earliest and broadest B cell restricted antigen is the CD19 antigen. The receptor for CD19 is an important functional regulator of normal and malignant B cell proliferation. It is expressed in all B cell precursor leukemias.

Specifications

Specifications

CD19
Monoclonal
APC
CD19
Purified
CD19
White blood cells.
Protein A/G
RUO
Antibody
Monoclonal
Human
Flow Cytometry
1G9
PBS with 0.2% carrier protein and 0.08% sodium azide
P15391
AW495831, B-lymphocyte antigen CD19, differentiation antigen CD19
Murine
IgG1
100 Tests
Store at 4°C
Primary
930
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit