Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD19 Rat anti-Mouse, AF 488, Clone: 6D5, Southern Biotech™
Rat Monoclonal Antibody
Supplier: Southern Biotechnology Associates 157530
Description
SouthernBiotech offers a variety of anti-mouse primary antibodies which are available in numerous formats including Alexa Fluor and Pacific Blue.
Specifications
CD19 | |
Monoclonal | |
0.5mg/mL | |
Supplied in PBS/NaN3 | |
Mouse CD19-expressing K562 human erythroleukemia cells | |
RUO | |
Mouse | |
IgG2a |
Flow Cytometry, Immunohistochemistry (Frozen) | |
6D5 | |
Alexa Fluor 488 | |
Rat | |
0.1 mg | |
Primary | |
Store at 2-8°C; Protect from light |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction