Learn More
CD19 Mouse anti-Human, V450, Clone: SJ25C1, BD
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 644491
Catalog No. BDB644491
Add to cart
Description
Description
The CD19 antibody, clone SJ25C1, is derived from the hybridization of Sp2/0 mouse myeloma cells with spleen cells isolated from BALB/c mice immunized with NALM1 and NALM16 cells.
The CD19 antibody recognizes a 90-kilodalton (kDa) antigen that is present on human B lymphocytes.
Specifications
Specifications
CD19
SJ25C1
V450
Conjugated
Human NALM1 + NALM16 cells
100 Tests
Immunology
Primary
Human
Flow Cytometry
25μg/mL
Phosphate buffered saline with gelatin and 0.1% sodium azide.
Mouse
IgG1, κ
RUO
Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed.
Monoclonal