Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD196 (CCR6), Mouse anti-Human, Clone: 11A9, BV711, BD

Manufacturer:  BD Biosciences 563923

Catalog No. BDB563923


Add to cart

Description

Description

Host Species: Mouse
Clone: 11A9
Isotype: IgG1, κ
Species Reactivity: Bb, Cyn, Hu, Rm
Immunogen: Human CD196/CCR6 Peptide

Flow cytometry

Specifications

Specifications

CD196
Affinity Purified
Human CD196/CCR6 Peptide
RUO
Monoclonal
11A9
Murine
50 Tests
Primary
Baboon, Cynomolgus, Human, Rhesus
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit