Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CD197 (CCR7); Clone: 150503; BD

Manufacturer:  BD Biosciences 563921

Catalog No. BDB563921

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Host Species: Mouse
Clone: 150503
Isotype: IgG2a
Species Reactivity: Hu
Immunogen: Human CCR7 Transfected Cell Line

Flow cytometry

Specifications

Specifications

CD197
Affinity Purified
Human CCR7 Transfected Cell Line
RUO
Monoclonal
150503
Murine
50 Tests
Primary
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit