Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD1a Mouse anti-Non-human primate, Clone: NA1/34-HLK, Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA133801
Description
MA1-33801 detects CD1a from Cynomolgus monkey samples. MA1-33801 has been successfully used in Flow cytometry and immunohistochemistry (frozen tissue) applications. Use 10 µL of the suggested working dilution to label 10^6 cells in 100 µL. The MA1-33801 immunogen is human thymocytes.
CD1a is a non-polymorphic MHC Class 1 related cell surface glycoprotein, expressed in association with Beta 2 microglobulin. CD1a is expressed by cortical thymocytes, Langerhan′s cells and by interdigitating cells. CD1a is also expressed by some maligcies of T cell lineage and in histiocytosis X. CD1a is expressed on cortical thymocytes, epidermal Langerhans cells, dendritic cells, on certain T-cell leukemias, and in various other tissues. CD1a is structurally related to the major histocompatibility complex (MHC) proteins and form heterodimers with beta-2-microglobulin. The CD1 proteins mediate the presentation of primarily lipid and glycolipid antigens of self or microbial origin to T cells. The human genome contains five CD1 family genes organized in a cluster on chromosome 1. The CD1 family members are thought to differ in their cellular localization and specificity for particular lipid ligands. CD1a localizes to the plasma membrane and recycles vesicles of the early endocytic system. At least five CD1 genes (CD1a, b, c, d, and e) are identified. Alternatively spliced transcript variants of CD1a have been observed, but their biological validity have not been determined.Specifications
CD1a | |
Monoclonal | |
1 mg/mL | |
CD1A | |
CD1, FCB6, HTA1, R4, T6, CD1A antigen, a polypeptide, T-cell surface antigen T6/Leu-6, T-cell surface glycoprotein CD1a, cluster of differentiation 1 A, cortical thymocyte antigen CD1A | |
Mouse | |
Ion-exchange chromatography | |
RUO | |
909 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Immunohistochemistry (Frozen) | |
NA1/34-HLK | |
Unconjugated | |
P06126 | |
CD1A | |
Human thymocytes | |
100 μg | |
Primary | |
Non-human Primate | |
Antibody | |
IgG2a |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title