Learn More
CD1c Mouse anti-Human, Alexa Fluor 647, Clone: F10/21A3, BD
Mouse Monoclonal Antibody
$122.00 - $316.00
Specifications
Antigen | CD1c |
---|---|
Clone | F10/21A3 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | Alexa Fluor 647 |
Description
The F10/21A3 monoclonal antibody specifically binds to CD1c. The CD1 family of transmembrane glycoproteins are structurally related to the classical major histocompatibility complex (MHC) proteins. CD1c is a type I transmembrane glycoprotein that forms heterodimers with beta-2-microglobulin. CD1c presents lipids and glycolipids of self or microbial origin to T cells. CD1c is expressed by Langerhans cells, dendritic cells, monocytes, cortical thymocytes, T cells, and some B cells.
Specifications
CD1c | |
Flow Cytometry | |
Alexa Fluor 647 | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide | |
IgG1 κ | |
Affinity Purified | |
CD1; R7; M241; BDCA1 |
F10/21A3 | |
Monoclonal | |
Mouse | |
Human | |
GM-CSF- and IL-4-activated Human Monocytes | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.