Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD1c Mouse anti-Human, BB515, Clone: F10/21A3, BD

Mouse Monoclonal Antibody

$127.30 - $282.50

Specifications

Antigen CD1c
Isotype IgG1, κ
Immunogen GM-CSF- and IL-4-activated Human Monocytes
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Conjugate BB515
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB565055
View Documents
BD Biosciences
565055
25 Tests
Each for $127.30
Add to cart
 
BDB565054
View Documents
BD Biosciences
565054
100 Tests
Each for $282.50
Add to cart
 
Description

Description

The F10/21A3 monoclonal antibody specifically binds to CD1c. The CD1 family of transmembrane glycoproteins are structurally related to the classical major histocompatibility complex (MHC) proteins. CD1c is a type I transmembrane glycoprotein that forms heterodimers with beta-2-microglobulin. CD1c presents lipids and glycolipids of self or microbial origin to T cells. CD1c is expressed by Langerhans cells, dendritic cells, monocytes, cortical thymocytes, T cells, and some B cells.

The antibody was conjugated to BD Horizon BB515 which was developed exclusively by BD Biosciences. With an excitation max of 490nm and an emission max of 515nm, BD Horizon BB515 can be excited by the 488nm laser and detected in a standard FITC set (e.g. 530/30-nm filter). This dye provides a much brighter alternative to FITC with less spillover into the PE detector.

Specifications

Specifications

CD1c
GM-CSF- and IL-4-activated Human Monocytes
BB515
Flow Cytometry
RUO
F10/21A3
CD1; R7; M241; BDCA1
IgG1, κ
Aqueous buffered solution containing ≤0.09% sodium azide.
Primary
Mouse
Affinity purified
Monoclonal
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
SDS
Product Certifications

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit