Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD1c Mouse anti-Human, PE, Clone: F10/21A3, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 564900

Catalog No. BDB564900


Add to cart

Description

Description

The F10/21A3 monoclonal antibody specifically binds to CD1c. The CD1 family of transmembrane glycoproteins are structurally related to the classical major histocompatibility complex (MHC) proteins. CD1c is a type I transmembrane glycoprotein that forms heterodimers with beta-2-microglobulin. CD1c presents lipids and glycolipids of self or microbial origin to T cells. CD1c is expressed by Langerhans cells, dendritic cells, monocytes, cortical thymocytes, T cells, and some B cells.

Specifications

Specifications

CD1c
F10/21A3
CD1; R7; M241; BDCA1
Affinity Purified
GM-CSF- and IL-4-activated Human Monocytes
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography.
RUO
Primary
Human
Flow Cytometry
PE
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Murine
IgG1, κ
100 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit