Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD2 Mouse anti-Human, BB515, Clone: RPA-2.10, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 565264

Catalog No. BDB565264


Add to cart

Description

Description

The RPA-2.10 monoclonal antibody specifically binds to CD2. CD2 is a 50 kDa single-chain transmembrane glycoprotein, also known as LFA-2 or the receptor for sheep erythrocytes. CD2 belongs to the immunoglobulin superfamily of proteins along with its ligand LFA-3, CD58. It is present on about 80-90% of human peripheral blood lymphocytes, greater than 95% of thymocytes, all T lymphocytes that form E-rosettes and a subset of NK cells. CD2 plays a role in T-cell signaling and in lymphocyte adhesion.

The antibody was conjugated to BD Horizon BB515 which was developed exclusively by BD Biosciences. With an excitation max of 490nm and an emission max of 515nm, BD Horizon BB515 can be excited by the 488nm laser and detected in a standard FITC set (e.g. 530/30-nm filter). This dye provides a much brighter alternative to FITC with less spillover into the PE detector.

Specifications

Specifications

CD2
RPA-2.10
LFA-2; Lymphocyte function antigen-2; T11/Leu-5; Rosette receptor
Affinity purified
Human Phytohemagglutinin-treated Lymphoblasts
50 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
BB515
Aqueous buffered solution containing ≤0.09% sodium azide
Murine
IgG1, κ
RUO
Primary
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit