Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD2 Mouse anti-Human, BUV395, Clone: RPA-2.10, BD

Mouse Monoclonal Antibody

$211.15 - $396.55

Specifications

Antigen CD2
Isotype IgG1, κ
Immunogen Human Phytohemagglutinin-treated Lymphoblast
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Conjugate Brilliant Ultraviolet 395
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB563820
View Documents
BD Biosciences
563820
25 Tests Each for $211.15
Add to cart
 
BDB563819
View Documents
BD Biosciences
563819
100 Tests Each for $396.55
Add to cart
 
Description

Description

The RPA-2.10 monoclonal antibody specifically binds to CD2. CD2 is a 50 kDa single-chain transmembrane glycoprotein, also known as LFA-2 or the receptor for sheep erythrocytes. CD2 belongs to the immunoglobulin superfamily of proteins along with its ligand LFA-3, CD58. It is present on about 80-90% of human peripheral blood lymphocytes, greater than 95% of thymocytes, all T lymphocytes that form E-rosettes and a subset of NK cells. CD2 plays a role in T-cell signaling and in lymphocyte adhesion.

The antibody was conjugated to BD Horizon™ BUV395 which has been exclusively developed by BD Biosciences as an optimal dye for use on a 355nm laser equipped instrument. With an Ex Max at 348nm and an Em Max at 395nm, this dye has virtually no spillover into any other detector. BD Horizon™ BUV395 can be excited with a 355nm laser and detected with a 379/28 filter.

Specifications

Specifications

CD2
Human Phytohemagglutinin-treated Lymphoblast
Brilliant Ultraviolet 395
Flow Cytometry
RUO
RPA-2.10
LFA-2; Lymphocyte function antigen-2; T11/Leu-5; Rosette receptor
IgG1, κ
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Primary
Murine
Affinity Purified
Monoclonal
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit