Learn More
CD2 Mouse anti-Human, V450, Clone: S5.2, BD
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 644485
Catalog No. BDB644485
Add to cart
Description
Description
CD2, clone S5.2, is derived from hybridization of mouse Sp2/0 myeloma cells with spleen cells from BALB/c mice immunized with T lymphocytes activated by mixed lymphocyte culture.
CD2 recognizes a human lymphocyte antigen, 45 to 50 kilodaltons (kDa), which also forms the binding site for sheep erythrocytes.
Specifications
Specifications
CD2
S5.2
V450
Conjugated
T lymphocytes activated by mix lymphocyte culture
100 Tests
Immunology
Primary
Human
Flow Cytometry
50μg/mL
Phosphate buffered saline with gelatin and 0.1% sodium azide.
Mouse
IgG2a, κ
RUO
Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed.
Monoclonal