Learn More
CD20 Mouse anti-Human, APC-H7, Clone: L27, BD
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 641396
Catalog No. BDB641396
Add to cart
Description
Description
CD20 clone L27 is derived from the hybridization of mouse Sp2/0 myeloma cells with spleen cells from BALB/c mice immunized with the LB lymphoblastoid cell line.
The CD20 antibody recognizes a phosphoprotein with a molecular weight of 35 or 37 kilodaltons (kDa), depending on the degree of phosphorylation. The antigen is not glycosylated.
Specifications
Specifications
CD20
L27
APC-H7
Affinity Purified
Human Lymphoblastoid Cell Line
100 Tests
Immunology
Primary
Human
Flow Cytometry
100μg/mL
Phosphate buffered saline with gelatin and 0.1% sodium azide.
Mouse
IgG1, κ
RUO
Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed.
Monoclonal