Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD20 Mouse anti-Human, BB515, Clone: 2H7, BD

Mouse Monoclonal Antibody

$99.00 - $220.00

Specifications

Antigen CD20
Isotype IgG2b, κ
Immunogen Human 6.16c1.3 B cell line
Formulation Aqueous buffered solution containing ≤0.09% sodium azide
Conjugate BB515
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB564569
View Documents
BD Biosciences
564569
25 Tests Each for $99.00
Add to cart
 
BDB564568
View Documents
BD Biosciences
564568
100 Tests Each for $220.00
Add to cart
 
Description

Description

The 2H7 monoclonal antibody specifically binds to CD20 that is encoded by the MS4A1 (Membrane-spanning 4-domains, subfamily A, member 1) gene. CD20 is a 33-37 kDa unglycosylated four-transmembrane phosphoprotein. CD20 is expressed on pre-B-cells, resting and activated B cells and follicular dendritic cells but not on plasma cells. Low level CD20 expression is observed on a small subset of normal circulating T lymphocytes. The CD20 molecule is involved in the regulation of B-cell activation.

Specifications

Specifications

CD20
Human 6.16c1.3 B cell line
BB515
Flow Cytometry
RUO
2H7
MS4A1; B1; Bp35; LEU-16; S7
IgG2b, κ
Aqueous buffered solution containing ≤0.09% sodium azide
Primary
Murine
Affinity purified
Monoclonal
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit