Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD200 Monoclonal Antibody (325531), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA523912
Description
In sandwich ELISAs, no cross-reactivity or interference with recombinant mouse CD200 is observed. Reconstitute at 0.5 mg/mL in sterile PBS. Endoxin level is <0.01 EU per 1 μg of the antibody by the LAL method.
CD200 Monoclonal antibody specifically detects CD200 in Human samples. It is validated for ELISA, Neutralization.Specifications
CD200 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P41217 | |
Cd200 | |
Mouse myeloma cell line NS0-derived recombinant human CD200 Gln31-Gly232 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Neutralization | |
325531 | |
Unconjugated | |
Cd200 | |
antigen identified by monoclonal antibody MRC OX-2; CD200; Cd200 antigen; Cd200 molecule; cell surface protein (thymocyte antigen identified by monoclonal antibody MRC-OX2); Cspmo2; hypothetical protein LOC534910; MOX1; MOX2; MRC; MRC OX-2 antigen; MRCOX2; My033; OX2; OX-2; OX-2 membrane glycoprotein | |
Mouse | |
Protein A/G | |
RUO | |
4345 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction