Learn More
CD201 Rat anti-Human, Brilliant Violet 510, Clone: RCR-252, BD
Rat Monoclonal Antibody
Manufacturer:  BD Biosciences 563624
Catalog No. BDB563624
Add to cart
Description
Description
The RCR-252 monoclonal antibody specifically binds to the endothelial cell protein C receptor (EPCR), which is also known as CD201. CD201 is a 25 kDa, type I transmembrane glycoprotein, expressed on endothelial cells in arteries, veins and capillaries. CD201 binds activated protein C, a serine protease that is involved in regulating blood coagulation. EPCR is reportedly expressed on several malignant cell lines. Malignant cell lines that express EPCR can show significant levels of protein C activation and this activation can be inhibited by anti-EPCR antibody
The antibody was conjugated to BD Horizon™ BV510 which is part of the BD Horizon Brilliant Violet™ family of dyes. With an Ex Max of 405-nm and Em Max at 510-nm, BD Horizon BV510 can be excited by the violet laser and detected in the BD Horizon V500 (525/50-nm) filter set. BD Horizon BV510 conjugates are useful for the detection of dim markers off the violet laser.
Specifications
Specifications
CD201
RCR-252
EPCR; Activated protein C receptor; APC receptor; PROCR; centrocyclin
Affinity Purified
EPCR-positive RE-1 Cells
100 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
Brilliant Violet 510
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Rat
IgG1, κ
RUO
Primary
Human