Learn More
EPCR Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA512519
Catalog No. PIPA512519
Add to cart
Description
Description
PROCR (CD201) is a receptor for activated protein C, a serine protease activated by and involved in the blood coagulation pathway. The protein is an N-glycosylated type I membrane protein that enhances the activation of protein C. Mutations in its gene have been associated with venous thromboembolism and myocardial infarction, as well as with late fetal loss during pregcy.
Specifications
Specifications
EPCR
Polyclonal
Unconjugated
PROCR
Liquid
PROCR
KLH conjugated synthetic peptide between 104-132 amino acids from the central region of human CD201
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
2 mg/mL
PBS with 0.09% sodium azide
Q9UNN8
Endothelial protein C receptor; Endothelial cell protein C receptor; Activated protein C receptor; APC receptor; CD201; EPCR; PROCR
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10544