Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Langerin (CD207) Mouse anti-Human, Unlabeled, Clone: 2G3 (also known as AB5_8.2G3.6 ), BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 564789

Catalog No. BDB564789


Add to cart

Description

Description

The 2G3 monoclonal antibody specifically binds to Langerin which is also known as CD207 or C-type lectin domain family 4 member K (CLEC4K). CD207 is a 48 kDa type II transmembrane glycoprotein that belongs to the C-type lectin family. CD207 is selectively expressed by Langerhans cells, an immature dendritic cell type that is found in the epidermis and mucosal epithelia, and by some other dendritic cell subsets. CD207 is expressed on the cell surface membrane and by intracellular Birbeck granules whose formation depends on CD207. CD207 binds to certain surface carbohydrates that are expressed by microbes. It appears to play a role in the uptake of these microbes for subsequent processing and microbial antigen presentation to T cells.

Specifications

Specifications

Langerin (CD207)
2G3 (also known as AB5_8.2G3.6 )
Unconjugated
Aqueous buffered solution containing ≤0.09% sodium azide.
Murine
IgG1, λ
0.1mg
Store undiluted at 4°C
Monoclonal
Flow Cytometry
0.5mg/mL
CLC4K; CLEC4K; C-type lectin domain family 4 member K
Affinity Purified
Human Langerin Recombinant Protein
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography.
RUO
Primary
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit