Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD21 Mouse anti-Human, PE, Clone: 544408, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA523542

Catalog No. PIMA523542


Add to cart

Description

Description

CD21 (complement receptor 2, CR2, C3D receptor, EBV receptor) binds C3 complement fragments, especially its breakdown fragments, which remain covalently attached to complement activating surfaces or antigen. CD21 has important roles in uptake and retention of immunocomplexes, survival of memory B cells and in development and maintece of the humoral response to T-dependent antigens. CD21 also serves as a key receptor for Epstein-Barr virus binding and is involved in targeting prions to follicular dendritic cells and expediting neuroinvasion following peripheral exposure to prions. A soluble form of the CD21 (sCD21) is shed from the lymphocyte surface and retains its ability to bind respective ligands. CD21 functions as receptor for C3d, C3dg and iC3b complement components, for EBV and for IFNalpha. CD21 binds to CD23 and associates with CD19, CD81 and Leu13 to form a large signal-transduction complex involved in B cell activation. Genetic variations in the CD21 gene are associated with susceptibility to systemic lupus erythematosus type 9 (SLEB9). Alternatively, spliced transcript variants encoding different isoforms of CD21 have been found.
Specifications

Specifications

CD21
Monoclonal
PE
CR2
Liquid
CR2
CHO-derived recombit human CD21 Ile21-Arg971
Protein A/G
RUO
Antibody
Monoclonal
Human
Flow Cytometry
544408
Buffered saline solution with BSA and 0.09% sodium azide
P20023
EBV receptor, complement C3d receptor, complement receptor type 2, epstein-Barr virus receptor
Mouse
IgG1
100 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
1380
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit