Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD22 Monoclonal Antibody (ICO-12), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA17649
Description
MA1-7649 detects CD22 from human samples. MA1-7649 has been successfully used in flow cytometry procedures. The MA1-7649 immunogen is B-lymphocytes.
CD22 Monoclonal antibody specifically detects CD22 in Human samples. It is validated for Flow Cytometry.Specifications
CD22 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.2% gelatin, 0.5% saccharose and 0.1% sodium azide; pH 7.2 | |
P20273 | |
CD22 | |
B-lymphocytes. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry | |
ICO-12 | |
Unconjugated | |
CD22 | |
A530093D23; B-cell receptor CD22; BL-CAM; B-lymphocyte cell adhesion molecule; B-lymphocyte cell adhesion molecule (BL-CAM); Cd22; CD22 antigen; CD22 molecule; FLJ22814; Lectin 2; Leu-14; Lyb8; Lyb-8; MGC130020; sialic acid binding Ig-like lectin 2; sialic acid-binding Ig-like lectin 2; Sialic acid-binding Ig-like lectin 2 (Siglec-2); Siglec2; Siglec-2; T-cell surface antigen Leu-14 | |
Mouse | |
Protein A | |
RUO | |
933 | |
4°C short term, -80°C long term | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction