Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD22 Mouse anti-Human, PE, Clone: 5.8HK, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA112062

Catalog No. 01-670-785

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

CD22 (BL-CAM) is a type 1 integral membrane glycoprotein with molecular weight of 130 to 140 kDa. CD22 is expressed in both the cytoplasm and cell membrane of B-lymphocytes. CD22 antigen appears early in B-cell lymphocyte differentiation at approximately the same stage as the CD19 antigen. Unlike other B-cell markers, CD22 membrane expression is limited to the late differentiation stages comprised between mature B cells (CD22+) and plasma cells (CD22-), and may thus prove useful in phenotyping mature leukemias. CD22 is also strongly expressed in hairy cell leukemia. CD22 preferentially binds to alpha2,6-linked sialic acid. The sialic acid recognition site can be masked by cis interactions with sialic acids on the same cell surface. Upon ligand induced tyrosine phosphorylation in the immune response seems to be involved in regulation of B cell antigen receptor signaling. CD22 plays a role in positive regulation through interaction with Src family tyrosine kinases and may also act as an inhibitory receptor by recruiting cytoplasmic phosphatases via their SH2 domains that block signal transduction through dephosphorylation of signaling molecules. CD22 is also strongly expressed in hairy cell leukemia. CD22 is also positive in diffuse large B-cell lymphoma and nodular lymphocyte predomice Hodgkin’s lymphoma, but negative in classical Hodgkin’s lymphoma.
Specifications

Specifications

CD22
Monoclonal
PE
CD22
Liquid
CD22
White blood cells.
Protein A/G
RUO
Antibody
Monoclonal
Human
Flow Cytometry
5.8HK
PBS with 0.2% carrier protein and 0.08% sodium azide
P20273
FLJ22814, MGC130020, SIGLEC-2, SIGLEC2, B-cell receptor CD22, B-lymphocyte cell adhesion molecule, BL-CAM, CD22 antigen, T-cell surface antigen Leu-14, sialic acid binding Ig-like lectin 2, sialic acid-binding Ig-like lectin 2
Mouse
IgG2b
100 Tests
4° C, do not freeze
Primary
933
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit