Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD220 Mouse, Unlabeled, Clone: 46, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610110

 View more versions of this product

Catalog No. BDB610110


Add to cart

Description

Description

Insulin Receptor (IR) is a transmembrane receptor tyrosine kinase which, upon insulin binding, initiates a cascade of events, including autophosphorylation, phosphorylation of cellular protein substrates, glucose transport, and glycogen synthesis. IR is synthesized as a large glycosylated precursor that is cleaved upon maturation into a 130kDa α-subunit with kinase activity and a 95kDa β-subunit. The active Insulin Receptor is a heterotetramer of homologus α and β subunits joined by disulfide bonds. Among the major cytosolic substrates of the Insulin Receptor are IRS-1 and -2, β-Adrenergic receptor, and pp15 (adipocyte lipid-binding protein, ALBP). Autophosphorylation of the IR recruits IRS-1 and -2 to the phosphotyrosines. Subsequently, the phosphorylated IRS-1 and -2 act as docking sites for other signaling proteins like PI3-Kinase, Shc, PTP1D, Nck, etc. In addition, the phosphatase LAR is tightly associated with the IR and LAR becomes activated after insulin stimulation dephosphorylating the IR and its substrates. Therefore, LAR provides a turn-off mechanism in insulin signaling.

Host Species: Mouse
Clone: 46
Isotype: IgG1
Species Reactivity: Mouse
Immunogen: Human Insulin pro-receptor β subunit 1006-1144
Formula Weight [Chemical]: 95kDa

Immunofluorescence, Immunoprecipitation, Western Blotting

Specifications

Specifications

CD220
46
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Western Blot
250μg/mL
Insulin Receptor β
Affinity Purified
Human Insulin pro-receptor β subunit 1006-1144
150μg
Immunology
Primary
Murine, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.