Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD23 Mouse anti-Human, PE, Clone: 138628, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA523549

Catalog No. PIMA523549


Add to cart

Description

Description

CD23 is a 45 kDa glycoprotein which is present on a subpopulation of freshly isolated peripheral blood and tonsil B cells and strongly expressed on EBV-transformed B lymphoblasts. The CD23 molecule is identical to the low affinity IgE receptor found on B cells. Expression of CD23 has been detected in neoplastic cells from cases of B cell chronic lymphocyctic leukaemia and some cases of centroblastic/centrocytic lymphoma. CD23 is present on a subpopulation of freshly isolated peripheral blood and tonsil B cells and strongly expressed on EBV-transformed B lymphoblasts. Functionally, CD23 is involved in B cell growth and differentiation, and IgE production. Further, CD23 has a soluble form that is a potent mitogenic factor. Diseases associated with CD23 dysfunction include chronic conjunctivitis and chronic lymphocytic leukemia.
Specifications

Specifications

CD23
Monoclonal
PE
FCER2
Liquid
FCER2
Human CD23
Protein A/G
RUO
Antibody
Monoclonal
Human
Flow Cytometry
138628
Buffered saline solution with BSA and 0.1% sodium azide
P06734
CD23, CD23A, CLEC4J, FCE2, IGEBF, BLAST-2, C-type lectin domain family 4, member J, CD23 antigen, fc-epsilon-RII, immunoglobulin E-binding factor, low affinity immunoglobulin epsilon Fc receptor, lymphocyte IgE receptor
Mouse
IgG1
100 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
2208
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit