Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD235a Mouse anti-Human, BB515, Clone: GA-R2 (HIR2), BD

Mouse Monoclonal Antibody

$116.30 - $288.00

Specifications

Antigen CD235a
Isotype IgG2b, κ
Formulation Aqueous buffered solution containing ≤0.09% sodium azide
Conjugate BB515
Primary or Secondary Primary
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB565234
View Documents
BD Biosciences
565234
25μG
Each for $116.30
Add to cart
 
BDB565233
View Documents
BD Biosciences
565233
0.1mg
Each for $288.00
Add to cart
 
Description

Description

The GA-R2 (also known as HIR2) monoclonal antibody specifically binds to CD235a and CD235b. CD235a is also known as Glycophorin A (GYPA, GPA, GLPA), Sialoglycoprotein alpha, MN sialoglycoprotein, or PAS-2. CD235b is otherwise known as Glycophorin B (GYPB, GPB, GLPB), Sialoglycoprotein delta, SS-active sialoglycoprotein, or PAS-3. CD235a and CD235b are type I transmembrane sialoglycoproteins that are expressed on human erythrocytes, erythroid precursor cells and certain leukemic cell types. CD235a carries blood group M and N antigens, whereas CD235b contains S, s, and U antigens. This antibody is useful for the identification and characterization of erythrocytes, certain myeloid leukemic cell types, and studies of erythroid cell development and infectious diseases with erythrocyte involvement. Glycophorins may play a role in preventing cell agglutination.

The antibody was conjugated to BD Horizon BB515 which was developed exclusively by BD Biosciences. With an excitation max of 490nm and an emission max of 515nm, BD Horizon BB515 can be excited by the 488nm laser and detected in a standard FITC set (e.g. 530/30-nm filter). This dye provides a much brighter alternative to FITC with less spillover into the PE detector.

Specifications

Specifications

CD235a
Aqueous buffered solution containing ≤0.09% sodium azide
Primary
Mouse
Affinity purified
Monoclonal
CD235ab; CD235a, Glycophorin-A, GYPA, GPA; CD235b; Glycophorin-B, GYPB, GPB
IgG2b, κ
BB515
Flow Cytometry
RUO
GA-R2 (HIR2)
0.2mg/mL
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
SDS
Product Certifications

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit