Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD249 Mouse anti-Human, Unlabeled, Clone: 2D3/APA, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 564532

Catalog No. BDB564532


Add to cart

Description

Description

The 2D3 monoclonal antibody specifically binds to CD249. CD249 is encoded by ENPEP and is also known as Aminopeptidase A (APA/AP-A), Glutamyl aminopeptidase (EAP), and gp160. CD249 is expressed by epithelial and endothelial cells in a variety of tissues including those that comprise the proximal tubules and glomeruli of the kidneys. It is also expressed by early B-lineage cells and cell lines. CD249 belongs to the peptidase M1 family and catalyzes the release of N-terminal glutamate (and to a lesser degree, aspartate) from peptides. This ectoenzyme catalyzes the conversion of angiotensin II to angiotensin III which is important in the local regulation of blood pressure. It may also play roles in the regulation of angiogenesis and development of B cells.

Specifications

Specifications

CD249
2D3/APA
Unconjugated
Aqueous buffered solution containing ≤0.09% sodium azide.
Murine
IgG1, κ
0.1mg
Store undiluted at 4°C
Monoclonal
Flow Cytometry
0.5mg/mL
EAP, Aminopeptidase A, Glutamyl aminopeptidase, APA, AP-A, gp160, ENPEP
Affinity Purified
Human Aminopeptidase A Transfected Cell Line
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography.
RUO
Primary
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit