Learn More
anti-CD25/IL-2 R alpha, PE, Polyclonal, R&D Systems™
Manufacturer:  R&D Systems FAB6750P100
 View more versions of this product
Catalog No. FAB6750P100
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
CD25/IL-2 R alpha
PE
Q544I2
Goat
IgG
100 Tests
Primary
25704
Murine, Rat
Flow Cytometry
Supplied in a saline solution containing BSA and Sodium Azide.
Affinity Purified
Mouse myeloma cell line NS0-derived recombinant mouse IL-2 Rα, Glu22-Lys236
Antigen Affinity-purified
RUO
Polyclonal
Detects mouse IL-2 Rα in ELISAs and Western blots. In sandwich ELISAs, less than 0.1% cross-reactivity with recombinant human IL-2 Rα is observed.