Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD265/RANK Rat anti-Mouse, Clone: R12-31, MilliporeSigma™
Rat Monoclonal Antibody
Supplier: MilliporeSigma 041507
Description
Anti-CD265/RANK Antibody, clone R12-31 detects level of CD265/RANK and has been published and validated for use in flow cytometry.Specifications
CD265/RANK | |
Monoclonal | |
Unconjugated | |
NP_033425.3 | |
Rat | |
Protein G purfied | |
Primary | |
2-8°C for one year | |
IgG2a κ |
Flow Cytometry | |
R12-31 | |
Purified rat monoclonal IgG2aκ in buffer containing 0.1MTris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide. | |
TNFRSF11A, RANK, OFE, ODFR, CD265, TRANCER | |
Mouse CD265/RANK-transfected 2PK3 cells. | |
100 μg | |
Mouse | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only