Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD27 Mouse anti-Human, Brilliant Violet 650, Clone: L128, BD

Mouse Monoclonal Antibody

$201.00 - $406.00

Specifications

Antigen CD27
Isotype IgG1
Immunogen Human Activated Peripheral Blood Cells
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Conjugate Brilliant Violet 650
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB563229
View Documents Promotion Details
BD Biosciences
563229
25 Tests
Each for $201.00
Add to cart
 
BDB563228
View Documents Promotion Details
BD Biosciences
563228
100 Tests
Each for $406.00
Add to cart
 
Description

Description

The L128 monoclonal antibody specifically binds to human CD27. CD27 is a 55-kDa disulfide-linked dimer that is a member of the nerve growth factor (NGF) super family. This family also includes CD40, rat OX40, tumor necrosis factor (TNF) receptors and CD95 (Fas). With its ligand CD70, CD27 acts in a co-stimulatory fashion on T lymphocytes. Present on most peripheral blood T lymphocytes and medullary thymocytes, the CD27 antigen is upregulated upon activation with the release of a soluble form, 28 to 32 kDa. It is also detected on a subpopulation of approximately 33% of circulating B lymphocytes. Following exposure to antigens, CD45RA+ T lymphocytes respond by upregulating the CD27 antigen. After maximal stimulation, the CD27 antigen cannot be re-expressed on long-term cultures or on CD45RA-CD27+ T lymphocytes. The CD4+CD27- population is contained within the memory CD45RO+ subset that proliferates after exposure to allergens. Two subpopulations of B lymphocytes bearing the CD27 antigen secrete IgM ( δ +) and IgG ( δ -).

Specifications

Specifications

CD27
Human Activated Peripheral Blood Cells
Brilliant Violet 650
Flow Cytometry
RUO
L128
TNFRSF7; Tumor necrosis factor receptor superfamily, member 7; Tp55; S152
IgG1
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Primary
Mouse
Affinity Purified
Monoclonal
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
SDS
Product Certifications

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit