Learn More
CD27 Hamster anti-Mouse, Brilliant Violet 510, Clone: LG.3A10, BD
Hamster Monoclonal Antibody
Supplier: BD Biosciences 563605
Description
The LG.3A10 monoclonal antibody specifically binds to CD27, a lymphocyte-restricted member of the Tumor Necrosis Factor Receptor family which binds to CD70. The CD27 molecule is a 45-kDa transmembrane glycoprotein which is constitutively expressed by lymphocytes of the T lineage: virtually all thymocytes and over 90% of peripheral T cells bearing both αβ and γδ T-cell receptors. CD27 cooperates with the pre-TCR in mediating thymocyte differentiation and expansion. In addition, one to ten percent of mature peripheral B cells express CD27, and CD27’s role in the differentiation of human plasma cells has been studied. Mouse NK cells, freshly isolated and IL-2-activated, also express CD27. In the bone marrow, CD27 is found on a progenitor population which provides short-term hematopoietic reconstitution. Cells of the myeloid lineage do not express CD27. Cross-linked LG.3A10 mAb has been reported to amplify the proliferative response of purified T lymphocytes to suboptimal mitogenic stimulation1 and to enhance NK-cell proliferation and IFN-γ production. In contrast, non-cross-linked LG.3A10 mAb inhibits CD3-induced pre-T cell development by interfering with the receptor-ligand interaction. This hamster mAb to a mouse leukocyte antigen has been observed to cross-react with a similar population of rat leukocytes.
Specifications
CD27 | |
Monoclonal | |
0.2mg/mL | |
Tnfrsf7; Tumor necrosis factor receptor superfamily member 7; Tp55; S152 | |
Hamster | |
Affinity Purified | |
RUO | |
Mouse | |
IgG1 κ |
Flow Cytometry | |
LG.3A10 | |
Brilliant Violet 510 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Armenian hamster fibroblast line ARHO12 transfected with mouse Cd27 cDNA | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.