Learn More
CD275 Mouse anti-Human, Alexa Fluor 647, Clone: 2D3/B7-H2, BD
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 564278
Catalog No. BDB564278
Add to cart
Description
Description
The 2D3 monoclonal antibody specifically binds to CD275, which is also known as B7 homolog 2 (B7-H2/B7H2) and B7-related protein 1 (B7RP-1). CD275 is a type I transmembrane glycoprotein that belongs to the Ig superfamily. CD275 is likewise known as Inducible T-cell co-stimulator ligand (ICOS Ligand /ICOSLG/ICOS-L). It can bind to CD278 (ICOS) and costimulate the proliferation and cytokine production of antigen-stimulated human T cells. CD275 is variable expressed by subsets of B cells, monocytes, macrophages, dendritic cells and endothelial cells.
Specifications
Specifications
CD275
2D3/B7-H2
ICOSLG; ICOS ligand; LICOS; B7-H2; B7 homolog 2; B7RP-1; GL50
Affinity Purified
Human B7-H2
100 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
Alexa Fluor 647
Aqueous buffered solution containing BSA and ≤0.09% sodium azide
Murine
IgG2b, κ
RUO
Primary
Human